Läs mer
N-Des(5-methylpyrazinecarbonyl)-N-ethylcarboxyl Glipizide, TRC
Produktkod. 30363362
Change view
Kvantitet:
500 mg
Produktkod. Kvantitet
30363362 500 mg
1 options
Produktkod. 30363362 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD292595500MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material N-Des(5-methylpyrazinecarbonyl)-N-ethylcarboxyl Glipizide
CAS 13554-93-3
Molekylformel C18H27N3O5S
Kvantitet 500 mg
Synonym [p-[(Cyclohexylcarbamoyl)sulfamoyl]phenethyl]carbamic Acid Ethyl Ester, [p-[(3-Cyclohexylureido)sulfonyl]phenethyl]carbamic Acid Ethyl Ester, Glipizide EP Impurity C
InChI formel InChI=1S/C18H27N3O5S/c1-2-26-18(23)19-13-12-14-8-10-16(11-9-14)27(24,25)21-17(22)20-15-6-4-3-5-7-15/h8-11,15H,2-7,12-13H2,1H3,(H,19,23)(H2,20,21,22)
LEDER CCOC(=O)NCCC1=CC=C(C=C1)S(=O)(=O)NC(=O)NC2CCCCC2
Molekylvikt (g/mol) 397.49
Formel vikt 397.1671