Läs mer
N-Des-t-boc-10-deacetyl-7,10-O-bis{[(2,2,2-trichloroethyl)oxy]carbonyl} Docetaxel, TRC
Produktkod. 30372043
Change view
Kvantitet:
1 mg
Produktkod. Kvantitet
30372043 1 mg
1 options
Produktkod. 30372043 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD4221101MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material N-Des-t-boc-10-deacetyl-7,10-O-bis{[(2,2,2-trichloroethyl)ox
CAS 114915-16-1
Molekylformel C44 H47 Cl6 N O16
Kvantitet 1 mg
Synonym Benzenepropanoic acid, β-amino-α-hydroxy-, (2aR,4S,4aS,6R,9S,11S,12S,12aR,12bS)-12b-(acetyloxy)-12-(benzoyloxy)-2a,3,4,4a,5,6,9,10,11,12,12a,12b-dodecahydro-11-hydroxy-4a,8,13,13-tetramethyl-5-oxo-4,6-bis[[(2,2,2-trichloroethoxy)carbonyl]oxy]-7,11-methano-1H-cyclodeca[3,4]benz[1,2-b]oxet-9-yl ester, (αR,βS)-, Benzenepropanoic acid, β-amino-α-hydroxy-, 12b-(acetyloxy)-12-(benzoyloxy)-2a,3,4,4a,5,6,9,10,11,12,12a,12b-dodecahydro-11-hydroxy-4a,8,13,13-tetramethyl-5-oxo-4,6-bis[[(2,2,2-trichloroethoxy)carbonyl]oxy]-7,11-methano-1H-cyclodeca[3,4]benz[1,2-b]oxet-9-yl ester, [2aR-[2aα,4β,4aβ,6β,9α(αR*,βS*),11α,12α,12aα,12bα]]-, 7,11-Methano-1H-cyclodeca[3,4]benz[1,2-b]oxete, benzenepropanoic acid deriv.
InChI formel InChI=1S/C44H47Cl6NO16/c1-21-25(63-36(56)30(53)29(51)23-12-8-6-9-13-23)17-42(59)34(66-35(55)24-14-10-7-11-15-24)32-40(5,33(54)31(28(21)39(42,3)4)65-38(58)62-20-44(48,49)50)26(64-37(57)61-19-43(45,46)47)16-27-41(32,18-60-27)67-22(2)52/h6-15,25-27,29-32,34,53,59H,16-20,51H2,1-5H3/t25-,26-,27+,29-,30+,31+,32-,34-,40+,41-,42+/m0/s1
LEDER CC(=O)O[C@@]12CO[C@@H]1C[C@H](OC(=O)OCC(Cl)(Cl)Cl)[C@]3(C)[C@@H]2[C@H](OC(=O)c4ccccc4)[C@]5(O)C[C@H](OC(=O)[C@H](O)[C@@H](N)c6ccccc6)C(=C([C@@H](OC(=O)OCC(Cl)(Cl)Cl)C3=O)C5(C)C)C
Molekylvikt (g/mol) 1058.56
Formel vikt 1055.1026