Läs mer
N-Des((tetrahydrofuran-2-yl)methanone)) Terazosin Dihydrochloride, TRC
Produktkod. 30357594
Change view
Kvantitet:
10 mg
Produktkod. Kvantitet
30357594 10 mg
1 options
Produktkod. 30357594 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD29794010MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material N-Des((tetrahydrofuran-2-yl)methanone) Terazosin Dihydrochloride
CAS 60548-08-5
Molekylformel C14H19N5O2.2[HCl]
Kvantitet 10 mg
Synonym 6,7-Dimethoxy-2-(1-piperazinyl)-4-quinazolinamine Dihydrochloride, 2-(1-Piperazinyl)-4-amino-6,7-dimethoxyquinazoline-2HCl, Terazosin Related Compound A (USP), Prazosin Impurity
InChI formel InChI=1S/C14H19N5O2.2ClH/c1-20-11-7-9-10(8-12(11)21-2)17-14(18-13(9)15)19-5-3-16-4-6-19;;/h7-8,16H,3-6H2,1-2H3,(H2,15,17,18);2*1H
LEDER Cl.Cl.COc1cc2nc(nc(N)c2cc1OC)N3CCNCC3
Molekylvikt (g/mol) 362.25
Formel vikt 361.1072