Läs mer
N-Desacetyl-N-formyl Thiocolchicoside, TRC
Produktkod. 30008186
Change view
Kvantitet:
1 mg
10 mg
25 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30008186 1 mg
30005144 25 mg
30003586 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30008186 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD2884101MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 219547-29-2
Molekylformel C26 H31 N O10 S
Molekylvikt (g/mol) 549.59
Synonym Formamide, N-[(7S)-3-(β-D-glucopyranosyloxy)-5,6,7,9-tetrahydro-1,2-dimethoxy-10-(methylthio)-9-oxobenzo[a]heptalen-7-yl]-, N-[(7S)-3-(β-D-Glucopyranosyloxy)-5,6,7,9-tetrahydro-1,2-dimethoxy-10-(methylthio)-9-oxobenzo[a]heptalen-7-yl]formamide, N-Desacetyl-N-formyl Thiocolchicoside, N-Deacetyl-N-formylthiocolchicoside, N-[(7S,12aRa)-3-(β-D-Glucopyranosyloxy)-1,2-dimethoxy-10-(methylsulfanyl)-9-oxo-5,6,7,9-tetrahydrobenzo[a]heptalen-7-yl]formamide
IUPAC-namn N-[(7S)-1,2-dimethoxy-10-methylsulfanyl-9-oxo-3-[(2S,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxy-6,7-dihydro-5H-benzo[a]heptalen-7-yl]formamide
LEDER COc1c(O[C@@H]2O[C@H](CO)[C@@H](O)[C@H](O)[C@H]2O)cc3CC[C@H](NC=O)C4=CC(=O)C(=CC=C4c3c1OC)SC
Specifikationer
Kemiskt namn eller material N-Desacetyl-N-formyl thiocolchicoside
Anteckningar om renhetsgrad HPLC
Kvantitet 1 mg
InChI formel InChI=1S/C26H31NO10S/c1-34-24-17(36-26-23(33)22(32)21(31)18(10-28)37-26)8-12-4-6-15(27-11-29)14-9-16(30)19(38-3)7-5-13(14)20(12)25(24)35-2/h5,7-9,11,15,18,21-23,26,28,31-33H,4,6,10H2,1-3H3,(H,27,29)/t15-,18+,21+,22-,23+,26+/m0/s1
Formel vikt 549.1669
Procent renhet >95%