Läs mer
N-Desbenzyl N-tert-butyloxycarbonyl Donepezil, TRC
Product Code. 30351308
Change view
Click to view available options
Kvantitet:
5 mg
Product Code. Kvantitet
30351308 5 mg
1 options
Product Code. 30351308 Supplier Toronto Research Chemicals Supplier No. TRCD2883955MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Kemiskt namn eller material N-Desbenzyl N-tert-butyloxycarbonyl Donepezil
CAS 948550-60-5
Molekylformel C22H31NO5
Kvantitet 5 mg
Synonym 4-[(2,3-Dihydro-5,6-dimethoxy-1-oxo-1H-inden-2-yl)methyl]-1-piperidinecarboxylic Acid 1,1-Dimethylethyl Ester
InChI formel InChI=1S/C22H31NO5/c1-22(2,3)28-21(25)23-8-6-14(7-9-23)10-16-11-15-12-18(26-4)19(27-5)13-17(15)20(16)24/h12-14,16H,6-11H2,1-5H3
LEDER COc1cc2CC(CC3CCN(CC3)C(=O)OC(C)(C)C)C(=O)c2cc1OC
IUPAC-namn tert-butyl 4-[(5,6-dimethoxy-3-oxo-1,2-dihydroinden-2-yl)methyl]piperidine-1-carboxylate
Molekylvikt (g/mol) 389.49
Formel vikt 389.2202