Läs mer
N-Descarboxymethyl-N-carboxyethyl Lacosamide (Impurity), TRC
Product Code. 30363456
Change view
Click to view available options
Kvantitet:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Kvantitet
30363456 10 mg
30357449 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 30363456 Supplier Toronto Research Chemicals Supplier No. TRCD28939010MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 1318777-56-8
Molekylformel C14 H20 N2 O3
Molekylvikt (g/mol) 264.32
Synonym N-Descarboxymethyl-N-carboxyethyl Lacosamide (Impurity), Propanamide, 3-methoxy-2-[(1-oxopropyl)amino]-N-(phenylmethyl)-, (2R)- (ACI), (2R)-3-Methoxy-2-[(1-oxopropyl)amino]-N-(phenylmethyl)propanamide (ACI), (R)-N-Benzyl-3-methoxy-2-propionamidopropanamide, (R)-N-benzyl-2-((1-hydroxypropylidene)amino)-3-methoxypropanimidic acid, N-Descarboxymethyl-N-carboxyethyl Lacosamide (Impurity)
IUPAC-namn (2R)-N-benzyl-3-methoxy-2-(propanoylamino)propanamide
LEDER CCC(=O)N[C@H](COC)C(=O)NCc1ccccc1
Specifications
Kemiskt namn eller material (2R)-3-Methoxy-2-[(1-oxopropyl)amino]-N-(phenylmethyl)propanamide
Kvantitet 10 mg
InChI formel InChI=1S/C14H20N2O3/c1-3-13(17)16-12(10-19-2)14(18)15-9-11-7-5-4-6-8-11/h4-8,12H,3,9-10H2,1-2H3,(H,15,18)(H,16,17)/t12-/m1/s1
Formel vikt 264.147