Läs mer
N-Descyanomethyl-N-acetamide Flonicamid, TRC
Specifikationer
Specifikationer
| Kemiskt namn eller material | Flonicamid-acetamide |
| CAS | 158062-96-5 |
| Molekylformel | C9 H8 F3 N3 O2 |
| Kvantitet | 100 mg |
| Synonym | N-(2-amino-2-oxoethyl)-4-(trifluoromethyl)pyridine-3-carboxamide, N-(2-Amino-2-oxoethyl)-4-(trifluoromethyl)-3-pyridinecarboxamide, N-(4-trifluoromethylnicotinoyl)glycinamide, TFNG-AM, Flonicamid-N-descyanomethyl-N-acetamide, Flonicamid-acetamide, N-Descyanomethyl-N-acetamide Flonicamid, N-(2-amino-2-oxoethyl)-4-(trifluoromethyl)pyridine-3-carboxamide, N-(2-Amino-2-oxoethyl)-4-(trifluoromethyl)-3-pyridinecarboxamide, N-(4-trifluoromethylnicotinoyl)glycinamide, TFNG-AM, Flonicamid-N-descyanomethyl-N-acetamide, Flonicamid-acetamide |
| InChI formel | InChI=1S/C9H8F3N3O2/c10-9(11,12)6-1-2-14-3-5(6)8(17)15-4-7(13)16/h1-3H,4H2,(H2,13,16)(H,15,17) |
| LEDER | NC(=O)CNC(=O)c1cnccc1C(F)(F)F |
| IUPAC-namn | N-(2-amino-2-oxoethyl)-4-(trifluoromethyl)pyridine-3-carboxamide |
| Molekylvikt (g/mol) | 247.174 |
| Formel vikt | 247.057 |