Läs mer
N-Desethyl Etifoxine, TRC
Produktkod. 30135394
Change view
Kvantitet:
25 mg
250 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30135394 250 mg
30006305 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30135394 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD289370250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 21715-43-5
Molekylformel C15 H13 Cl N2 O
Molekylvikt (g/mol) 272.73
IUPAC-namn 6-chloro-4-methyl-4-phenyl-3,1-benzoxazin-2-amine
LEDER CC1(OC(=Nc2ccc(Cl)cc12)N)c3ccccc3
Specifikationer
Kemiskt namn eller material N-Desethyl Etifoxine
Analyt- eller komponentnamn N-Desethyl Etifoxine
Frakt skick Room Temperature
InChI formel InChI=1S/C15H13ClN2O/c1-15(10-5-3-2-4-6-10)12-9-11(16)7-8-13(12)18-14(17)19-15/h2-9H,1H3,(H2,17,18)
Lösningstyp Neat
Industry Type Pharmaceutical
Kvantitet 250 mg
Formel vikt 272.0716