Läs mer
N-Desethyl-N-propyl Oxybutynin, TRC
Produktkod. 17985095
Change view
Kvantitet:
5 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
17985095 50 mg
17922295 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 17985095 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD28950050MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 1215677-72-7
Molekylformel C23 H33 N O3
Molekylvikt (g/mol) 371.51
Synonym 4-(Ethylpropylamino)but-2-ynyl (RS)-2-Cyclohexyl-2-hydroxy-2-phenylacetate, Oxybutynin Hydrochloride Imp. E (EP), Ethylpropyl Analogue of Oxybutynin, Benzeneacetic acid, α-cyclohexyl-α-hydroxy-, 4-(ethylpropylamino)-2-butyn-1-yl ester (ACI), 4-(Ethylpropylamino)-2-butyn-1-yl α-cyclohexyl-α-hydroxybenzeneacetate (ACI)
IUPAC-namn 4-[ethyl(propyl)amino]but-2-ynyl 2-cyclohexyl-2-hydroxy-2-phenylacetate
LEDER CCCN(CC)CC#CCOC(=O)C(O)(C1CCCCC1)c2ccccc2
Specifikationer
Kemiskt namn eller material 4-(Ethylpropylamino)but-2-ynyl (RS)-2-Cyclohexyl-2-hydroxy-2-phenylacetate
Kvantitet 50 mg
InChI formel InChI=1S/C23H33NO3/c1-3-17-24(4-2)18-11-12-19-27-22(25)23(26,20-13-7-5-8-14-20)21-15-9-6-10-16-21/h5,7-8,13-14,21,26H,3-4,6,9-10,15-19H2,1-2H3
Formel vikt 371.246