Läs mer
N-Desethyl Vardenafil, TRC
Produktkod. 30134471
Change view
Kvantitet:
5 mg
10 mg
50 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30134471 10 mg
30012642 50 mg
17997544 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30134471 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD28995010MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 448184-46-1
Molekylformel C21 H28 N6 O4 S
Molekylvikt (g/mol) 460.55
Synonym Imidazo[5,1-f][1,2,4]triazin-4(1H)-one, 2-[2-ethoxy-5-(1-piperazinylsulfonyl)phenyl]-5-methyl-7-propyl-, Piperazine, 1-[[3-(1,4-dihydro-5-methyl-4-oxo-7-propylimidazo[5,1-f][1,2,4]triazin-2-yl)-4-ethoxyphenyl]sulfonyl]- (9CI), N-Desethylvardenafil, 2-[2-Ethoxy-5-(piperazin-1-ylsulfonyl)phenyl]-5-methyl-7-propylimidazo[5,1-f][1,2,4]triazin-4(3H)-one
IUPAC-namn 2-(2-ethoxy-5-piperazin-1-ylsulfonylphenyl)-5-methyl-7-propyl-3H-imidazo[5,1-f][1,2,4]triazin-4-one
LEDER CCCc1nc(C)c2C(=O)NC(=Nn12)c3cc(ccc3OCC)S(=O)(=O)N4CCNCC4
Specifikationer
Kemiskt namn eller material N-Desethylvardenafil
Anteckningar om renhetsgrad HPLC
Kvantitet 10 mg
InChI formel InChI=1S/C21H28N6O4S/c1-4-6-18-23-14(3)19-21(28)24-20(25-27(18)19)16-13-15(7-8-17(16)31-5-2)32(29,30)26-11-9-22-10-12-26/h7-8,13,22H,4-6,9-12H2,1-3H3,(H,24,25,28)
Formel vikt 460.1893
Procent renhet >95%