Läs mer
N-Deshydroxy Ciclopirox, TRC
Produktkod. 30137387
Change view
Kvantitet:
10 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30137387 100 mg
30008805 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30137387 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD289995100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 67587-24-0
Molekylformel C12 H17 N O
Molekylvikt (g/mol) 191.27
Synonym 6-Cyclohexyl-4-methylpyridin-2(1H)-one, Ciclopirox Olamine Imp. C (EP), 2(1H)-Pyridinone, 6-cyclohexyl-4-methyl- (9CI), 6-Cyclohexyl-4-methyl-2(1H)-pyridone, N-Deshydroxy Ciclopirox
IUPAC-namn 6-cyclohexyl-4-methyl-1H-pyridin-2-one
LEDER CC1=CC(=O)NC(=C1)C2CCCCC2
Specifikationer
Kemiskt namn eller material 6-Cyclohexyl-4-methylpyridin-2(1H)-one
Anteckningar om renhetsgrad HPLC
Kvantitet 100 mg
InChI formel InChI=1S/C12H17NO/c1-9-7-11(13-12(14)8-9)10-5-3-2-4-6-10/h7-8,10H,2-6H2,1H3,(H,13,14)
Formel vikt 191.131
Procent renhet >95%