Läs mer
N-Desisopropyl N-Chlorophenylchlorguanide Hydrochloride, TRC
Produktkod. 30356188
Change view
Kvantitet:
500 mg
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30356188 5 g
30352699 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30356188 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD2903355G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 13590-98-2
Molekylformel C14 H13 Cl2 N5 . Cl H
Molekylvikt (g/mol) 358.65
Synonym N,N'-bis(4-Chlorophenyl)imidodicarbonimidic Diamide Hydrochloride (1:1), Biguanide, 1,5-bis(p-chlorophenyl)-, monohydrochloride (8CI), Imidodicarbonimidic diamide, N,N'-bis(4-chlorophenyl)-, monohydrochloride (9CI), ST 39008, 1,5-Bis-(4-Chlorophenyl)biguanide Hydrochloride, N1,N3-bis(4-Chlorophenyl)imidodicarbonimidic Diamide Hydrochloride
IUPAC-namn 1-(4-chlorophenyl)-3-[N-(4-chlorophenyl)carbamimidoyl]guanidine;hydrochloride
LEDER Cl.Clc1ccc(NC(=N)NC(=N)Nc2ccc(Cl)cc2)cc1
Specifikationer
Kemiskt namn eller material 1,5-bis(4-chlorophenyl)biguanide hydrochloride
Anteckningar om renhetsgrad HPLC
Kvantitet 5 g
InChI formel InChI=1S/C14H13Cl2N5.ClH/c15-9-1-5-11(6-2-9)19-13(17)21-14(18)20-12-7-3-10(16)4-8-12;/h1-8H,(H5,17,18,19,20,21);1H
Formel vikt 357.0315
Procent renhet >95%