Läs mer
N-Desmethyl 3-Hydroxy Mepivacaine, TRC
Produktkod. 30357746
Change view
Kvantitet:
1 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30357746 1 mg
30354232 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30357746 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD2918451MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 247061-17-2
Molekylformel C14H20N2O2
Molekylvikt (g/mol) 248.32
Synonym N-(3-Hydroxy-2,6-dimethylphenyl)-2-piperidinecarboxamide
LEDER CC1=C(C(=C(C=C1)O)C)N=C(C2CCCCN2)O
Specifikationer
Kemiskt namn eller material N-Desmethyl 3-Hydroxy Mepivacaine
Kvantitet 1 mg
InChI formel InChI=1S/C14H20N2O2/c1-9-6-7-12(17)10(2)13(9)16-14(18)11-5-3-4-8-15-11/h6-7,11,15,17H,3-5,8H2,1-2H3,(H,16,18)
Formel vikt 248.1525