Läs mer
N-Desmethyl-4-hydroxy Tamoxifen Beta-D-Glucuronide (E/Z Mixture), TRC
Produktkod. 30366631
Change view
Kvantitet:
10 mg
Produktkod. Kvantitet
30366631 10 mg
1 options
Produktkod. 30366631 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD29204210MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material N-Desmethyl-4-hydroxy Tamoxifen beta-D-Glucuronide (E,Z mixture)
CAS 152764-52-8
Molekylformel C31 H35 N O8
Kvantitet 10 mg
Synonym 4-[1-[4-[2-(Methylamino)ethoxy]phenyl]-2-phenyl-1-buten-1-yl]phenyl β-D-Glucopyranosiduronic Acid, (E/Z)-Endoxifen Glucuronide
InChI formel InChI=1S/C31H35NO8/c1-3-24(19-7-5-4-6-8-19)25(20-9-13-22(14-10-20)38-18-17-32-2)21-11-15-23(16-12-21)39-31-28(35)26(33)27(34)29(40-31)30(36)37/h4-16,26-29,31-35H,3,17-18H2,1-2H3,(H,36,37)/t26-,27-,28+,29-,31+/m0/s1
LEDER CCC(=C(c1ccc(OCCNC)cc1)c2ccc(O[C@@H]3O[C@@H]([C@@H](O)[C@H](O)[C@H]3O)C(=O)O)cc2)c4ccccc4
IUPAC-namn (2S,3S,4S,5R,6S)-3,4,5-trihydroxy-6-[4-[1-[4-[2-(methylamino)ethoxy]phenyl]-2-phenyl-but-1-enyl]phenoxy]tetrahydropyran-2-carboxylic acid
Molekylvikt (g/mol) 549.61
Formel vikt 549.2363