Läs mer
N-Desmethyl Asenapine Hydrochloride, TRC
Produktkod. 30361000
Change view
Kvantitet:
5 mg
Produktkod. Kvantitet
30361000 5 mg
1 options
Produktkod. 30361000 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD2907405MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material N-desmethyl Asenapine hydrochloride
CAS 1170701-78-6
Molekylformel C16 H14 Cl N O . Cl H
Kvantitet 5 mg
Synonym 1H-Dibenz[2,3:6,7]oxepino[4,5-c]pyrrole, 5-chloro-2,3,3a,12b-tetrahydro-, hydrochloride (1:1), (3aR,12bR)-rel- (ACI), rel-(3aR,12bR)-5-Chloro-2,3,3a,12b-tetrahydro-1H-dibenz[2,3:6,7]oxepino[4,5-c]pyrrole hydrochloride, N-Desmethylasenapine hydrochloride, trans-5-Chloro-2,3,3a,12b-tetrahydro-1H-dibenz[2,3:6,7]oxepino[4,5-c]pyrrole hydrochloride
InChI formel InChI=1S/C16H14ClNO.ClH/c17-10-5-6-16-12(7-10)14-9-18-8-13(14)11-3-1-2-4-15(11)19-16;/h1-7,13-14,18H,8-9H2;1H/t13-,14-;/m1./s1
LEDER Cl.Clc1ccc2Oc3ccccc3[C@H]4CNC[C@@H]4c2c1
Molekylvikt (g/mol) 308.2
Formel vikt 307.0531