Läs mer
N-Desmethyl Clobazam, TRC
Produktkod. 30362130
Change view
Kvantitet:
10 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30362130 10 mg
30354621 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30362130 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD29154010MG
Denna vara kräver en prisuppdatering innan den beställs.
Begär ett pris för denna vara här.
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material N-Desmethylclobazam
CAS 22316-55-8
Molekylformel C15 H11 Cl N2 O2
Kvantitet 10 mg
Synonym 8-chloro-1-phenyl-1H-1,5-benzodiazepine-2,4(3H,5H)-dione, 7-Chloro-5-phenyl-1,2,4,5-tetrahydro-3H-1,5-benzodiazepine-2,4-dione, Clofazin, Demethylclobazam, Desmethylclobazam, N-Demethylclobazam, N-Desmethylclobazam, Norclobazam, Clobazam Imp. A (EP)
InChI formel InChI=1S/C15H11ClN2O2/c16-10-6-7-12-13(8-10)18(11-4-2-1-3-5-11)15(20)9-14(19)17-12/h1-8H,9H2,(H,17,19)
LEDER Clc1ccc2NC(=O)CC(=O)N(c3ccccc3)c2c1
IUPAC-namn 7-chloro-5-phenyl-1H-1,5-benzodiazepine-2,4-dione
Molekylvikt (g/mol) 286.71
Formel vikt 286.0509