Läs mer
N-Desmethyl Diphenhydramine Hydrochloride, TRC
Produktkod. 30007324
Change view
Kvantitet:
100 mg
250 mg
500 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30007324 100 mg
30008062 500 mg
30010141 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30007324 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD291780100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 53499-40-4
Molekylformel C16 H19 N O . Cl H
Molekylvikt (g/mol) 277.79
Synonym 2-(Diphenylmethoxy)-N-methylethanamine Hydrochloride, Nordiphenhydramine Hydrochloride, Dimenhydrinate Imp. F (EP) as Hydrochloride, Diphenhydramine Imp. A Hydrochloride, Diphenhydramine Hydrochloride Imp. A (EP) as Hydrochloride
IUPAC-namn 2-benzhydryloxy-N-methylethanamine;hydrochloride
LEDER Cl.CNCCOC(c1ccccc1)c2ccccc2
Specifikationer
Kemiskt namn eller material Nordiphenhydramine Hydrochloride
Anteckningar om renhetsgrad HPLC
Kvantitet 100 mg
InChI formel InChI=1S/C16H19NO.ClH/c1-17-12-13-18-16(14-8-4-2-5-9-14)15-10-6-3-7-11-15;/h2-11,16-17H,12-13H2,1H3;1H
Formel vikt 277.1233
Procent renhet >95%