Läs mer
N-Desmethyl Eletriptan, TRC
Produktkod. 30365717
Change view
Kvantitet:
250 mg
Produktkod. Kvantitet
30365717 250 mg
1 options
Produktkod. 30365717 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD291666250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material N-Desmethyl Eletriptan
CAS 153525-55-4
Molekylformel C21 H24 N2 O2 S
Kvantitet 250 mg
Synonym N-Desmethyl Eletriptan, (R)-5-[2-(phenylsulfonyl)ethyl]-3-(2-pyrrolidinylmethyl)-1H-Indole, N-Demethyl Eletriptan
InChI formel InChI=1S/C21H24N2O2S/c24-26(25,19-6-2-1-3-7-19)12-10-16-8-9-21-20(13-16)17(15-23-21)14-18-5-4-11-22-18/h1-3,6-9,13,15,18,22-23H,4-5,10-12,14H2/t18-/m1/s1
LEDER O=S(=O)(CCc1ccc2[nH]cc(C[C@H]3CCCN3)c2c1)c4ccccc4
IUPAC-namn 5-[2-(benzenesulfonyl)ethyl]-3-[[(2R)-pyrrolidin-2-yl]methyl]-1H-indole
Molekylvikt (g/mol) 368.493
Formel vikt 368.156