Läs mer
N-Desmethyl Galanthamine, TRC
Produktkod. 30012478
Change view
Kvantitet:
5 mg
10 mg
25 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30012478 25 mg
30007000 5 mg
30009778 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30012478 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD29203525MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 41303-74-6
Molekylformel C16 H19 N O3
Molekylvikt (g/mol) 273.327
Synonym 6H-Benzofuro[3a,3,2-ef][2]benzazepin-6-ol, 4a,5,9,10,11,12-hexahydro-3-methoxy-, (4aS,6R,8aS)-, Galanthamine, 10-demethyl-, (4aS,6R,8aS)-4a,5,9,10,11,12-Hexahydro-3-methoxy-6H-benzofuro[3a,3,2-ef][2]benzazepin-6-ol, 6H-Benzofuro[3a,3,2-ef][2]benzazepin-6-
LEDER COc1ccc2CNCC[C@@]34C=C[C@H](O)C[C@@H]3Oc1c24
Specifikationer
Kemiskt namn eller material N-Desmethyl Galanthamine
Anteckningar om renhetsgrad HPLC
Kvantitet 25 mg
InChI formel InChI=1S/C16H19NO3/c1-19-12-3-2-10-9-17-7-6-16-5-4-11(18)8-13(16)20-15(12)14(10)16/h2-5,11,13,17-18H,6-9H2,1H3/t11-,13-,16-/m0/s1
Formel vikt 273.136
Procent renhet >95%