Läs mer
N-Desmethyl-N-cyclopentyl cis-ent-Tadalafil, TRC
Produktkod. 30337030
Change view
Kvantitet:
25 mg
250 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30337030 250 mg
30347223 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30337030 Leverantör Toronto Research Chemicals Leverantörsnummer TRCB199005250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material N-Desmethyl-N-cyclopentyl cis-ent-Tadalafil
CAS 171488-19-0
Molekylformel C26 H25 N3 O4
Kvantitet 250 mg
Synonym N-Desmethyl-N-cyclopentyl cis-ent-Tadalafil
InChI formel InChI=1S/C26H25N3O4/c30-23-13-28(16-5-1-2-6-16)26(31)20-12-18-17-7-3-4-8-19(17)27-24(18)25(29(20)23)15-9-10-21-22(11-15)33-14-32-21/h3-4,7-11,16,20,25,27H,1-2,5-6,12-14H2/t20-,25+/m0/s1
LEDER O=C1CN(C2CCCC2)C(=O)[C@@H]3Cc4c([nH]c5ccccc45)[C@H](N13)c6ccc7OCOc7c6
IUPAC-namn (2R,8S)-2-(1,3-benzodioxol-5-yl)-6-cyclopentyl-3,6,17-triazatetracyclo[8.7.0.0^{3,8}.0^{11,16}]heptadeca-1(10),11,13,15-tetraene-4,7-dione
Molekylvikt (g/mol) 443.494
Formel vikt 443.185