Läs mer
N-Desmethyl N-Ethyl Doxepin, TRC
Produktkod. 30360859
Change view
Kvantitet:
10 mg
Produktkod. Kvantitet
30360859 10 mg
1 options
Produktkod. 30360859 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD29143510MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material N-Desmethyl N-Ethyl Doxepin
CAS 70805-90-2
Molekylformel C20H23NO
Kvantitet 10 mg
Synonym 3-(Dibenz[b,e]oxepin-11(6H)-ylidene)-N-ethyl-N-methyl-1-propanamine, Dibenz[b,e]oxepin 1-Propanamine Deriv.
InChI formel InChI=1S/C20H23NO/c1-3-21(2)14-8-10-16-15-17-9-4-6-12-19(17)22-20-13-7-5-11-18(16)20/h4-7,9-13H,3,8,14-15H2,1-2H3/b16-10+
LEDER CCN(C)CC\C=C\1/Cc2ccccc2Oc3ccccc13
Molekylvikt (g/mol) 293.4
Formel vikt 293.178