Läs mer
N-Desmethyl N-Ethyl Olanzapine, TRC
Produktkod. 30366801
Change view
Kvantitet:
100 mg
Produktkod. Kvantitet
30366801 100 mg
1 options
Produktkod. 30366801 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD291675100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material N-Desmethyl N-Ethyl Olanzapine
CAS 1355034-54-6
Molekylformel C18H22N4S
Kvantitet 100 mg
Synonym 4-(4-Ethyl-1-piperazinyl)-2-methyl-5H-thienobenzodiazepine
InChI formel InChI=1S/C18H22N4S/c1-3-21-8-10-22(11-9-21)17-14-12-13(2)23-18(14)20-16-7-5-4-6-15(16)19-17/h4-7,12,20H,3,8-11H2,1-2H3
LEDER CCN1CCN(CC1)C2=Nc3ccccc3Nc4sc(C)cc24
Molekylvikt (g/mol) 326.46
Formel vikt 326.1565