Läs mer
N-Desmethyl,-N-nitroso-promethazine, TRC
Produktkod. 30365739
Change view
Kvantitet:
10 mg
Produktkod. Kvantitet
30365739 10 mg
1 options
Produktkod. 30365739 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD39395010MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material N-Desmethyl,-N-nitroso-promethazine
CAS 94511-44-1
Molekylformel C16H17N3OS
Kvantitet 10 mg
Synonym 10H-Phenothiazine-10-ethanamine, N,α-dimethyl-N-nitroso- (9CI, ACI), N,α-Dimethyl-N-nitroso-10H-phenothiazine-10-ethanamine (ACI), N-Nitroso-N-Desmethyl Promethazine
InChI formel InChI=1S/C16H17N3OS/c1-12(18(2)17-20)11-19-13-7-3-5-9-15(13)21-16-10-6-4-8-14(16)19/h3-10,12H,11H2,1-2H3
LEDER O=NN(C)C(C)CN1C=2C=CC=CC2SC=3C=CC=CC31
IUPAC-namn N-(1-(10H-phenothiazin-10-yl)propan-2-yl)-N-methylnitrous amide
Molekylvikt (g/mol) 299.39
Formel vikt 299.11