Läs mer
N-Desmethyl-N-nitrosolevofloxacine, Mikromol™
Produktkod. 30051556
Change view
Kvantitet:
25 mg
Produktkod. Kvantitet
30051556 25 mg
1 options
Produktkod. 30051556 Leverantör Mikromol Leverantörsnummer MM0846.150025
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
N-Desmethyl-N-nitrosolevofloxacine
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material N-Desmethyl-N-nitrosolevofloxacine
CAS 1152314-62-9
Analyt- eller komponentnamn N-Desmethyl-N-nitrosolevofloxacine
Frakt skick Ice pack (-20°C)
Molekylformel C17 H17 F N4 O5
Certifikat och efterlevnad ISO 9001
Synonym (3S)-9-Fluoro-2,3-dihydro-3-methyl-10-(4-nitroso-1-piperazinyl)-7-oxo-7H-pyrido[1,2,3-de]-1,4-benzoxazine-6-carboxylic Acid, 7H-Pyrido[1,2,3-de]-1,4-benzoxazine-6-carboxylic acid, 9-fluoro-2,3-dihydro-3-methyl-10-(4-nitroso-1-piperazinyl)-7-oxo-, (3S)- (ACI), (3S)-9-Fluoro-2,3-dihydro-3-methyl-10-(4-nitroso-1-piperazinyl)-7-oxo-7H-pyrido[1,2,3-de]-1,4-benzoxazine-6-carboxylic acid (ACI), N-Desmethyl-N-nitrosolevofloxacine
InChI formel InChI=1S/C17H17FN4O5/c1-9-8-27-16-13-10(15(23)11(17(24)25)7-22(9)13)6-12(18)14(16)20-2-4-21(19-26)5-3-20/h6-7,9H,2-5,8H2,1H3,(H,24,25)/t9-/m0/s1
LEDER C[C@H]1COc2c(N3CCN(CC3)N=O)c(F)cc4C(=O)C(=CN1c24)C(=O)O
Molekylvikt (g/mol) 376.3391
Kvantitet 25 mg
Formel vikt 376.1183 g/mol