Läs mer
N-Desmethyl Perazine Dimalonic Acid Salt, TRC
Specifikationer
Specifikationer
| Kemiskt namn eller material | N-Desmethyl Perazine Dimalonic Acid Salt |
| CAS | 60285-35-0 |
| Molekylformel | C19H23N3S . C6H8O8 |
| Kvantitet | 2.5 mg |
| Synonym | 10-[3-(1-Piperazinyl)propyl]-10H-phenothiazine Malonic Acid Salt, 10-(3-Piperazinylpropyl)phenothiazine Malonic Acid Salt, Demethylperazine Malonic Acid Salt, Desmethylperazine Malonic Acid Salt, Norperazine Malonic Acid Salt |
| InChI formel | InChI=1S/C19H23N3S/c1-3-8-18-16(6-1)22(17-7-2-4-9-19(17)23-18)13-5-12-21-14-10-20-11-15-21/h1-4,6-9,20H,5,10-15H2 |
| LEDER | C(CN1CCNCC1)CN2c3ccccc3Sc4ccccc24 |
| Molekylvikt (g/mol) | 325.5 |