Läs mer
N-Desmethyl Propamocarb Hydrochloride, TRC
Produktkod. 30354107
Change view
Kvantitet:
1 g
Produktkod. Kvantitet
30354107 1 g
1 options
Produktkod. 30354107 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD2924601G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Propamocarb-N-desmethyl hydrochloride
CAS 2287345-42-8
Molekylformel C8 H18 N2 O2 . Cl H
Kvantitet 1 g
Synonym Propamocarb-N-desmethyl Hydrochloride, Carbamic acid, N-[3-(methylamino)propyl]-, propyl ester, hydrochloride (1:1) (ACI)
InChI formel InChI=1S/C8H18N2O2.ClH/c1-3-7-12-8(11)10-6-4-5-9-2;/h9H,3-7H2,1-2H3,(H,10,11);1H
LEDER Cl.CCCOC(=O)NCCCNC
IUPAC-namn propyl N-[3-(methylamino)propyl]carbamate;hydrochloride
Molekylvikt (g/mol) 210.7
Formel vikt 210.1135