Läs mer
N-Desmethyl Regorafenib (Pyridine)-N-oxide, TRC
Produktkod. 30133595
Change view
Kvantitet:
10 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30133595 100 mg
30008905 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30133595 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD293280100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 835621-12-0
Molekylformel C20H13ClF4N4O4
Molekylvikt (g/mol) 484.79
Synonym 4-[4-[[[[4-Chloro-3-(trifluoromethyl)phenyl]amino]carbonyl]amino]-3-fluorophenoxy]-2-pyridinecarboxamide 1-Oxide, Regorafenib metabolite M5
LEDER O=C(NC1=CC=C(Cl)C(C(F)(F)F)=C1)NC2=C(F)C=C(OC3=CC=N(C(C(N)=O)=C3)=O)C=C2
Specifikationer
Kemiskt namn eller material N-Desmethyl Regorafenib (Pyridine)-N-oxide
Anteckningar om renhetsgrad HPLC
Kvantitet 100 mg
InChI formel InChI=1S/C20H13ClF4N4O4/c21-14-3-1-10(7-13(14)20(23,24)25)27-19(31)28-16-4-2-11(8-15(16)22)33-12-5-6-29(32)17(9-12)18(26)30/h1-9H,(H2,26,30)(H2,27,28,31)
Formel vikt 484.0561
Procent renhet >95%