Läs mer
N-Desmethyl Riociguat, TRC
Specifikationer
Specifikationer
| Kemiskt namn eller material | N-Desmethyl Riociguat |
| CAS | 625115-52-8 |
| Molekylformel | C19 H17 F N8 O2 |
| Kvantitet | 50 mg |
| Synonym | Carbamic acid, N-[4,6-diamino-2-[1-[(2-fluorophenyl)methyl]-1H-pyrazolo[3,4-b]pyridin-3-yl]-5-pyrimidinyl]-, methyl ester, Carbamic acid, [4,6-diamino-2-[1-[(2-fluorophenyl)methyl]-1H-pyrazolo[3,4-b]pyridin-3-yl]-5-pyrimidinyl]-, methyl ester (9CI), BAY 60-4552, Methyl [4,6-diamino-2-[1-(2-fluorobenzyl)-1H-pyrazolo[3,4-b]pyridin-3-yl]pyrimidin-5-yl]carbamate, Neliciguat, Nelociguat, N-Desmethyl Riociguat, N-Desmethylriociguat, Methyl [4,6-diamino-2-[1-[(2-fluorophenyl)methyl]-1H-pyrazolo[3,4-b]pyridin-3-yl]pyrimidin-5-yl]carbamate, Riociguat Imp. A (EP) |
| InChI formel | InChI=1S/C19H17FN8O2/c1-30-19(29)24-14-15(21)25-17(26-16(14)22)13-11-6-4-8-23-18(11)28(27-13)9-10-5-2-3-7-12(10)20/h2-8H,9H2,1H3,(H,24,29)(H4,21,22,25,26) |
| LEDER | COC(=O)Nc1c(N)nc(nc1N)c2nn(Cc3ccccc3F)c4ncccc24 |
| IUPAC-namn | methyl N-[4,6-diamino-2-[1-[(2-fluorophenyl)methyl]pyrazolo[3,4-b]pyridin-3-yl]pyrimidin-5-yl]carbamate |
| Molekylvikt (g/mol) | 408.39 |
| Formel vikt | 408.1459 |