Läs mer
N-Desmethyl Tadalafil, TRC
Produktkod. 30008212
Change view
Kvantitet:
5 mg
10 mg
25 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30008212 25 mg
17911765 5 mg
17952555 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30008212 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD29380025MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 171596-36-4
Molekylformel C21 H17 N3 O4
Molekylvikt (g/mol) 375.38
Synonym Pyrazino[1',2':1,6]pyrido[3,4-b]indole-1,4-dione, 6-(1,3-benzodioxol-5-yl)-2,3,6,7,12,12a-hexahydro-, (6R,12aR)-, Pyrazino[1',2':1,6]pyrido[3,4-b]indole-1,4-dione, 6-(1,3-benzodioxol-5-yl)-2,3,6,7,12,12a-hexahydro-, (6R-trans)-, Nortadalafil
IUPAC-namn (2R,8R)-2-(1,3-benzodioxol-5-yl)-3,6,17-triazatetracyclo[8.7.0.0^{3,8}.0^{11,16}]heptadeca-1(10),11,13,15-tetraene-4,7-dione
LEDER O=C1CNC(=O)[C@H]2Cc3c([nH]c4ccccc34)[C@H](N12)c5ccc6OCOc6c5
Specifikationer
Kemiskt namn eller material N-Desmethyl Tadalafil
Anteckningar om renhetsgrad HPLC
Kvantitet 25 mg
InChI formel InChI=1S/C21H17N3O4/c25-18-9-22-21(26)15-8-13-12-3-1-2-4-14(12)23-19(13)20(24(15)18)11-5-6-16-17(7-11)28-10-27-16/h1-7,15,20,23H,8-10H2,(H,22,26)/t15-,20-/m1/s1
Formel vikt 375.1219
Procent renhet >95%