Läs mer
N-Desmethyl Terbinafine Hydrochloride, TRC
Produktkod. 30352078
Change view
Kvantitet:
1 mg
Produktkod. Kvantitet
30352078 1 mg
1 options
Produktkod. 30352078 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD2943051MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material N-Demethylterbinafine Hydrochloride
CAS 152830-98-3
Molekylformel C20 H23 N . Cl H
Kvantitet 1 mg
Synonym N-Demethylterbinafine Hydrochloride
InChI formel InChI=1S/C20H23N.ClH/c1-20(2,3)14-7-4-8-15-21-16-18-12-9-11-17-10-5-6-13-19(17)18;/h4-6,8-13,21H,15-16H2,1-3H3;1H/b8-4+;
LEDER Cl.CC(C)(C)C#C\C=C\CNCc1cccc2ccccc12
IUPAC-namn (E)-6,6-dimethyl-N-(naphthalen-1-ylmethyl)hept-2-en-4-yn-1-amine;hydrochloride
Molekylvikt (g/mol) 313.86
Formel vikt 313.1597