Läs mer
N-Desmethyl Topotecan, TRC
Produktkod. 30349216
Change view
Kvantitet:
5 mg
Produktkod. Kvantitet
30349216 5 mg
1 options
Produktkod. 30349216 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD2944005MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material N-Desmethyl Topotecan
CAS 190710-79-3
Molekylformel C22 H21 N3 O5
Kvantitet 5 mg
Synonym 1H-Pyrano[3',4':6,7]indolizino[1,2-b]quinoline-3,14(4H,12H)-dione, 4-ethyl-4,9-dihydroxy-10-[(methylamino)methyl]-, (4S)-, 1H-Pyrano[3',4':6,7]indolizino[1,2-b]quinoline-3,14(4H,12H)-dione, 4-ethyl-4,9-dihydroxy-10-[(methylamino)methyl]-, (S)-, (4S)-4-Ethyl-4,9-dihydroxy-10-[(methylamino)methyl]-1H-pyrano[3',4':6,7]indolizino[1,2-b]quinoline-3,14(4H,12H)-dione, N-Desmethyl topotecan
InChI formel InChI=1S/C22H21N3O5/c1-3-22(29)15-7-17-19-11(9-25(17)20(27)14(15)10-30-21(22)28)6-12-13(8-23-2)18(26)5-4-16(12)24-19/h4-7,23,26,29H,3,8-10H2,1-2H3/t22-/m0/s1
LEDER CC[C@@]1(O)C(=O)OCC2=C1C=C3N(Cc4cc5c(CNC)c(O)ccc5nc34)C2=O
Molekylvikt (g/mol) 407.42
Formel vikt 407.1481