Läs mer
N-Desmethyl Zolmitriptan, TRC
Produktkod. 30008109
Change view
Kvantitet:
1 mg
10 mg
25 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30008109 1 mg
30005185 10 mg
30009182 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30008109 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD2967651MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 139264-35-0
Molekylformel C15 H19 N3 O2
Molekylvikt (g/mol) 273.33
Synonym (4S)-4-[[3-[2-(Methylamino)ethyl]-1H-indol-5-yl]methyl]-2-oxazolidinone, 2-Oxazolidinone, 4-[[3-[2-(methylamino)ethyl]-1H-indol-5-yl]methyl]-, (S)-, 183C91, Zolmitriptan Related Compound A (USP)
IUPAC-namn (4S)-4-[[3-[2-(methylamino)ethyl]-1H-indol-5-yl]methyl]-1,3-oxazolidin-2-one
LEDER CNCCc1c[nH]c2ccc(C[C@H]3COC(=O)N3)cc12
Specifikationer
Kemiskt namn eller material N-Desmethyl Zolmitriptan
Kvantitet 1 mg
InChI formel InChI=1S/C15H19N3O2/c1-16-5-4-11-8-17-14-3-2-10(7-13(11)14)6-12-9-20-15(19)18-12/h2-3,7-8,12,16-17H,4-6,9H2,1H3,(H,18,19)/t12-/m0/s1
Formel vikt 273.1477