Läs mer
N-Desmethylcarboxy Terbinafine, TRC
Produktkod. 30006927
Change view
Kvantitet:
1 mg
5 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30006927 5 mg
30012250 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30006927 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD2914055MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 99473-15-1
Molekylformel C20 H21 N O2
Molekylvikt (g/mol) 307.39
Synonym 5-Hepten-3-ynoic acid, 2,2-dimethyl-7-[(1-naphthalenylmethyl)amino]-, (5E)-, 5-Hepten-3-ynoic acid, 2,2-dimethyl-7-[(1-naphthalenylmethyl)amino]-, (E)-, (5E)-2,2-Dimethyl-7-[(1-naphthalenylmethyl)amino]-5-hepten-3-ynoic acid, N-Desmethylcarboxyterbinafine
IUPAC-namn (E)-2,2-dimethyl-7-(naphthalen-1-ylmethylamino)hept-5-en-3-ynoic acid
LEDER CC(C)(C#C\C=C\CNCc1cccc2ccccc12)C(=O)O
Specifikationer
Kemiskt namn eller material N-Desmethylcarboxy Terbinafine
Kvantitet 5 mg
InChI formel InChI=1S/C20H21NO2/c1-20(2,19(22)23)13-6-3-7-14-21-15-17-11-8-10-16-9-4-5-12-18(16)17/h3-5,7-12,21H,14-15H2,1-2H3,(H,22,23)/b7-3+
Formel vikt 307.1572