Läs mer
N-Desmethylcyclopropyl Mitapivat, TRC
Produktkod. 30353469
Change view
Kvantitet:
5 mg
Produktkod. Kvantitet
30353469 5 mg
1 options
Produktkod. 30353469 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD2998205MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material N-Desmethylcyclopropyl Mitapivat
CAS 1260081-87-5
Molekylformel C20H20N4O3S
Kvantitet 5 mg
Synonym Mitapivat Impurity, N-[4-(1-Piperazinylcarbonyl)phenyl]-8-quinolinesulfonamide
InChI formel InChI=1S/C20H20N4O3S/c25-20(24-13-11-21-12-14-24)16-6-8-17(9-7-16)23-28(26,27)18-5-1-3-15-4-2-10-22-19(15)18/h1-10,21,23H,11-14H2
LEDER O=C(C1=CC=C(C=C1)NS(=O)(=O)C2=CC=CC3=CC=CN=C32)N4CCNCC4
IUPAC-namn N-(4-(piperazine-1-carbonyl)phenyl)quinoline-8-sulfonamide
Molekylvikt (g/mol) 396.46
Formel vikt 396.13