Läs mer
N-Desmethylolanzapine, Lipomed
Produktkod. 30461013
Change view
Kvantitet:
100 mg
Produktkod. Kvantitet
30461013 100 mg
1 options
Produktkod. 30461013 Leverantör Lipomed Leverantörsnummer LPMOLZ1899FB100
Lipomed delivers certified reference materials you can trust as neats, native solutions and stable isotope labelled solutions, with precise documentation and expert support. From forensic to clinical labs, choose Lipomed for confidence, compliance, and analytical excellence.
N-Desmethylolanzapine
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Desmethylolanzapine
CAS 161696-76-0
Analyt- eller komponentnamn Desmethylolanzapine
Frakt skick Room Temperature
Molekylformel C16 H18 N4 S
Synonym N-Desmethylolanzapine, Desmethylolanzapine, 2-Methyl-4-(1-piperazinyl)-10 H-thieno[2,3-b][1,5 ]benzodiazepine, 4'-Desmethylolanzapine, LY 170055, N-Demethylolanzapine
InChI formel InChI=1 S/C16H21NO2.ClH/c1-12(2)17-10-14(18)11-19-16-9-5-7-13-6-3-4-8-15(13)16;/h3-9,12,14,17-18 H,10-11H2,1-2H3;1 H
LEDER Cc1cc2C(=Nc3ccccc3Nc2s1)N4CCNCC4
IUPAC-namn 2-methyl-4-piperazin-1-yl-10 H-thieno[2,3-b][1,5 ]benzodiazepine
Industry Type Pharmaceutical
Molekylvikt (g/mol) 298.41
Kvantitet 100 mg
Formel vikt 298.1252 g/mol