Läs mer
N-Desmethylolanzapine, Mikromol™
Produktkod. 30457876
Change view
Kvantitet:
25 mg
100 mg
4 x 25 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30457876 4 x 25 mg
30049167 25 mg
17943436 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30457876 Leverantör Mikromol Leverantörsnummer MM0329.0100254AMP
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
N-Desmethylolanzapine, Mikromol
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Desmethylolanzapine
CAS 161696-76-0
Analyt- eller komponentnamn Desmethylolanzapine
Frakt skick Room Temperature
Molekylformel C16 H18 N4 S
Certifikat och efterlevnad ISO 17025
Synonym N-Desmethylolanzapine, Desmethylolanzapine, 2-Methyl-4-(1-piperazinyl)-10H-thieno[2,3-b][1,5]benzodiazepine, 4'-Desmethylolanzapine, LY 170055, N-Demethylolanzapine
InChI formel InChI=1S/C16H18N4S/c1-11-10-12-15(20-8-6-17-7-9-20)18-13-4-2-3-5-14(13)19-16(12)21-11/h2-5,10,17,19H,6-9H2,1H3
Lösningstyp Neat
LEDER Cc1cc2C(=Nc3ccccc3Nc2s1)N4CCNCC4
IUPAC-namn 2-methyl-4-piperazin-1-yl-10H-thieno[2,3-b][1,5]benzodiazepine
Molekylvikt (g/mol) 298.41
Kvantitet 4 x 25 mg
Formel vikt 298.1252 g/mol