Läs mer
N-Despropyl Propafenone Oxalate Salt, TRC
Produktkod. 30137164
Change view
Kvantitet:
2.5 mg
10 mg
25 mg
Förpackningsstorlek:
2.5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet unitSize
30137164 25 mg -
30004895 10 mg -
30006780 2.5 mg 2.5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30137164 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD28866025MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 93885-20-2
Molekylformel C18 H21 N O3 . C2 H2 O4
Molekylvikt (g/mol) 389.4
Synonym 1-Propanone, 1-[2-(3-amino-2-hydroxypropoxy)phenyl]-3-phenyl-, ethanedioate (1:1) (salt) (9CI)
IUPAC-namn 1-[2-(3-amino-2-hydroxypropoxy)phenyl]-3-phenylpropan-1-one;oxalic acid
LEDER NCC(O)COc1ccccc1C(=O)CCc2ccccc2.OC(=O)C(=O)O
Specifikationer
Kemiskt namn eller material N-Depropyl Propafenone Oxalate Salt
Kvantitet 25 mg
InChI formel InChI=1S/C18H21NO3.C2H2O4/c19-12-15(20)13-22-18-9-5-4-8-16(18)17(21)11-10-14-6-2-1-3-7-14;3-1(4)2(5)6/h1-9,15,20H,10-13,19H2;(H,3,4)(H,5,6)
Formel vikt 389.1475