Läs mer
N-Despropyl Ropinirole Hydrochloride, TRC
Produktkod. 30363252
Change view
Kvantitet:
2.5 mg
Produktkod. Kvantitet
30363252 2.5 mg
1 options
Produktkod. 30363252 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD2973752.5MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material N-Despropyl Ropinirole Hydrochloride
CAS 173990-76-6
Molekylformel C13 H18 N2 O . Cl H
Kvantitet 2.5 mg
Synonym 2H-Indol-2-one, 1,3-dihydro-4-[2-(propylamino)ethyl]-, hydrochloride (1:1), 2H-Indol-2-one, 1,3-dihydro-4-[2-(propylamino)ethyl]-, monohydrochloride (9CI), 1,3-Dihydro-4-[2-(propylamino)ethyl]-2H-indol-2-one hydrochloride (1:1), 4-[2-(Propylamino)ethyl]-1,3-dihydro-2H-indol-2-one Hydrochloride, N-Despropyl Ropinirole
InChI formel InChI=1S/C13H18N2O.ClH/c1-2-7-14-8-6-10-4-3-5-12-11(10)9-13(16)15-12;/h3-5,14H,2,6-9H2,1H3,(H,15,16);1H
LEDER Cl.CCCNCCc1cccc2NC(=O)Cc12
IUPAC-namn 4-[2-(propylamino)ethyl]-1,3-dihydroindol-2-one;hydrochloride
Molekylvikt (g/mol) 254.76
Formel vikt 254.1186