Läs mer
N-Dodecyl-acetamide, TRC
Produktkod. 30370888
Change view
Kvantitet:
10 mg
50 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30370888 100 mg
30370551 50 mg
30378296 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30370888 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD562050100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 3886-80-4
Molekylformel C14H29NO
Molekylvikt (g/mol) 227.39
LEDER CCCCCCCCCCCCNC(=O)C
Specifikationer
Kemiskt namn eller material N-Dodecyl-acetamide
Kvantitet 100 mg
InChI formel InChI=1S/C14H29NO/c1-3-4-5-6-7-8-9-10-11-12-13-15-14(2)16/h3-13H2,1-2H3,(H,15,16)