Learn More
N-Dodecylpyridinium chloride, Sigma-Aldrich™
Specifications
Specifications
| Chemical Name or Material | N-Dodecylpyridinium chloride |
| CAS | 104-74-5 |
| Density | 1.04 g/cm³ at 20°C |
| Flash Point | 160°C |
| Melting Range | 87°C to 89°C |
| Molecular Formula | CH3(CH2)11C5H5NCl |
| Quantity | 25 kg |
| Synonym | N-Dodecylpyridinium chloride, N-Laurylpyridinium chloride, N-Dodecylpyridinium chloride |
| Molecular Weight (g/mol) | 283.88 |
| Grade | Not Available |