Läs mer
N-Ethoxycarbonyl Norastemizole, TRC
Produktkod. 30377564
Change view
Kvantitet:
100 mg
Produktkod. Kvantitet
30377564 100 mg
1 options
Produktkod. 30377564 Leverantör Toronto Research Chemicals Leverantörsnummer TRCE891545100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material N-Ethoxycarbonyl Norastemizole
CAS 84501-68-8
Molekylformel C22H25FN4O2
Kvantitet 100 mg
Synonym Ethyl 4-[[1-[(4-Fluorophenyl)methyl]-1h-benzimidazol-2-yl]amino]piperidine-1-carboxylate, 4-[[1-[(4-Fluorophenyl)methyl]-1H-benzimidazol-2-yl]amino]-1-piperidinecarboxylic acid Ethyl Ester
InChI formel InChI=1S/C22H25FN4O2/c1-2-29-22(28)26-13-11-18(12-14-26)24-21-25-19-5-3-4-6-20(19)27(21)15-16-7-9-17(23)10-8-16/h3-10,18H,2,11-15H2,1H3,(H,24,25)
LEDER CCOC(=O)N1CCC(CC1)Nc2nc3ccccc3n2Cc4ccc(F)cc4
Molekylvikt (g/mol) 396.46
Formel vikt 396.1962