missing translation for 'onlineSavingsMsg'
Learn More

N-Ethyl-2,5-difluorobenzamide, 97%, Thermo Scientific™

Catalog Number p-7058142
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
948717-61-1
C9H9F2NO
185.174
MFCD11843991
VMNRGEZTDCWQLU-UHFFFAOYSA-N
benzamide, 2,5-difluoro-n-ethyl
17683297
N-ethyl-2,5-difluorobenzamide
CCNC(=O)C1=C(C=CC(=C1)F)F
This item is not returnable. View return policy
948717-61-1
C9H9F2NO
185.174
MFCD11843991
VMNRGEZTDCWQLU-UHFFFAOYSA-N
benzamide, 2,5-difluoro-n-ethyl
17683297
N-ethyl-2,5-difluorobenzamide
CCNC(=O)C1=C(C=CC(=C1)F)F

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Quantity 250 mg
Formula Weight 185.17
Percent Purity 97%
Chemical Name or Material N-Ethyl-2,5-difluorobenzamide
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'