Läs mer
N-Ethyl-3-phenylpropan-1-amine Hydrochloride, Mikromol™
Produktkod. 30463763
Change view
Kvantitet:
25 mg
100 mg
4 x 25 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30463763 4 x 25 mg
30051416 25 mg
17972026 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30463763 Leverantör Mikromol Leverantörsnummer MM0454.0400254AMP
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
N-Ethyl-3-phenylpropan-1-amine Hydrochloride, Mikromol
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material N-Ethyl-3-phenylpropan-1-amine Hydrochloride
CAS 13125-63-8
Analyt- eller komponentnamn N-Ethyl-3-phenylpropan-1-amine Hydrochloride
Frakt skick Room Temperature
Molekylformel C11 H17 N . Cl H
Certifikat och efterlevnad ISO 17025
Synonym Alverine Citrate Imp. C (EP), Alverine Imp. C (EP), N-Ethyl-3-phenylpropan-1-amine Hydrochloride, Alverine Citrate Imp. C (EP) as Hydrochloride, Alverine Citrate Impurity C as Hydrochloride, Alverine Impurity C as Hydrochloride
InChI formel InChI=1S/C11H17N.ClH/c1-2-12-10-6-9-11-7-4-3-5-8-11;/h3-5,7-8,12H,2,6,9-10H2,1H3;1H
Lösningstyp Neat
LEDER Cl.CCNCCCc1ccccc1
IUPAC-namn N-ethyl-3-phenylpropan-1-amine;hydrochloride
Molekylvikt (g/mol) 199.72
Kvantitet 4 x 25 mg
Formel vikt 199.1128 g/mol