Läs mer
N-Ethyl-4-methylbenzenesulfonamide, 98%, Thermo Scientific™
Produktkod. 15408498
Change view
Kvantitet:
100 g
500 g
Förpackningsstorlek:
100g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet unitSize
15408498 100 g 100g
15418498 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 15408498 Leverantör Thermo Scientific Alfa Aesar Leverantörsnummer H61905.22
Request Bulk or Custom Format
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Se alternativa produkter
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 80-39-7
Molekylformel C9H13NO2S
Molekylvikt (g/mol) 199.268
MDL-nummer MFCD00048511
InChI-nyckel OHPZPBNDOVQJMH-UHFFFAOYSA-N
Synonym n-ethyl-p-toluenesulfonamide, santicizer 3, n-tosylethylamine, benzenesulfonamide, n-ethyl-4-methyl, n-ethyl-p-tolylsulfonamide, n-ethyl-4-toluenesulfonamide, p-tolueneethylsulfonamide, ethyl tosylamide, p-toluenesulfonyl-n-ethylamide, n-ethyl-p-methylbenzenesulfonamide
PubChem CID 6637
IUPAC-namn N-ethyl-4-methylbenzenesulfonamide
LEDER CCNS(=O)(=O)C1=CC=C(C=C1)C
Specifikationer
Kemiskt namn eller material N-Ethyl-4-methylbenzenesulfonamide
Smältpunkt 63°C to 65°C
Kokpunkt 208°C (745 mmHg)
Kvantitet 100 g
Formel vikt 199.27
Procent renhet 98%