Läs mer
N-Ethyl-N-methyl-3-vinylphenyl Carbamate, TRC
Produktkod. 30375619
Change view
Kvantitet:
250 mg
Produktkod. Kvantitet
30375619 250 mg
1 options
Produktkod. 30375619 Leverantör Toronto Research Chemicals Leverantörsnummer TRCE925205250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material N-Ethyl-N-methyl-3-vinylphenyl Carbamate
CAS 1346602-84-3
Molekylformel C12 H15 N O2
Kvantitet 250 mg
Synonym Carbamic acid, N-ethyl-N-methyl-, 3-ethenylphenyl ester, 3-Vinylphenyl ethyl(methyl)carbamate, N-Ethyl-N-methyl-3-vinylphenyl Carbamate, Rivastigmine Related Compound F
InChI formel InChI=1S/C12H15NO2/c1-4-10-7-6-8-11(9-10)15-12(14)13(3)5-2/h4,6-9H,1,5H2,2-3H3
LEDER CCN(C)C(=O)Oc1cccc(C=C)c1
IUPAC-namn (3-ethenylphenyl) N-ethyl-N-methylcarbamate
Molekylvikt (g/mol) 205.25
Formel vikt 205.1103