Läs mer
N-Ethyl-N-nitroso-o-toluidine, TRC
Produktkod. 30417133
Change view
Kvantitet:
500 mg
Produktkod. Kvantitet
30417133 500 mg
1 options
Produktkod. 30417133 Leverantör Toronto Research Chemicals Leverantörsnummer TRCN703685500MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material N-Nitroso N-Methyl-Phenethylamine
CAS 41030-87-9
Molekylformel C9H12N2O
Kvantitet 500 mg
Synonym Benzenamine','N-ethyl-2-methyl-N-nitroso- (9CI','ACI), N-Ethyl-2-methyl-N-nitrosobenzenamine (ACI), o-Toluidine','N-ethyl-N-nitroso- (7CI), N-Nitroso-N-ethyl-o-tolylamine, N-Ethyl-2-methyl-N-nitrosoaniline, N-Ethyl-N-nitroso-o-toluidine
InChI formel InChI=1S/C9H12N2O/c1-3-11(10-12)9-7-5-4-6-8(9)2/h4-7H,3H2,1-2H3
LEDER CCN(N=O)c1ccccc1C
IUPAC-namn N-ethyl-N-(2-methylphenyl)nitrous amide
Molekylvikt (g/mol) 164.2044
Formel vikt 164.095