Learn More
N-Ethylmaleimide, 98+%, Thermo Scientific Chemicals
Description
Cysteine alkylating agent N-Ethylmaleimide is a protein thiol modifier which inhibits apoptotic DNA fragmentation. It is a sulfhydryl reagent and finds application in experimental biochemical studies as well as in enzymology. It is involved in the modification of cysteine residues in proteins and peptides. It acts as a Michael acceptor and reacts with nucelophiles like thiols. It is employed in the inhibition of Mg2+-dependent internucleosomal DNA fragmentation.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Cysteine alkylating agent N-Ethylmaleimide is a protein thiol modifier which inhibits apoptotic DNA fragmentation. It is a sulfhydryl reagent and finds application in experimental biochemical studies as well as in enzymology. It is involved in the modification of cysteine residues in proteins and peptides. It acts as a Michael acceptor and reacts with nucelophiles like thiols. It is employed in the inhibition of Mg2+-dependent internucleosomal DNA fragmentation.
Solubility
Soluble in ethanol, water, chloroform, methanol and toluene.
Notes
Light sensitive. Store in cool place. Incompatible with strong oxidizing agents, strong acids and strong bases.
Specifications
Melting Point | 43°C to 46°C |
Boiling Point | 209°C to 211°C |
Flash Point | 73°C (163°F) |
Odor | Pungent |
Quantity | 10 g |
UN Number | UN2928 |
Beilstein | 112448 |
Sensitivity | Light sensitive |
Merck Index | 14,3822 |
Solubility Information | Soluble in ethanol,water,chloroform,methanol and toluene. |
Show More |
Safety and Handling
- N-Ethylmaleimide
- Danger
- Acute toxicity Category 2
- Skin corrosion/irritation Category 1 B
- Acute toxicity Category 4
- Skin sensitiser Category 1
- H300-Fatal if swallowed.
- H312-Harmful in contact with skin.
- H314-Causes severe skin burns and eye damage.
- H317-May cause an allergic skin reaction.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P405-Store locked up.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
RUO – Research Use Only