missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ N-Fmoc-D-alpha-phenylglycine, 98%

Brand:  Alfa Aesar™ H63823.06

Additional Details : CAS Number : 111524-95-9 Weight : 0.00500kg

Quantity 5g

 View more versions of this product

Product Code. 15450729

  • 983.72 SEK / 5g


Estimated Shipment date
from Supplier 17-12-2018
Add to basket

Specifications

Chemical Identifiers

111524-95-9
373.408
PCJHOCNJLMFYCV-OAQYLSRUSA-N
2756108
C1=CC=C(C=C1)C(C(=O)O)NC(=O)OCC2C3=CC=CC=C3C4=CC=CC=C24
C23H19NO4
MFCD00190891
fmoc-d-phg-oh, 2r-2-9h-fluoren-9-ylmethoxycarbonylamino-2-phenyl-acetic acid, r-2-9h-fluoren-9-yl methoxy carbonyl amino-2-phenylacetic acid, fmoc-d-phenylglycine, n-alpha-9-fluorenylmethyloxycarbonyl-d-phenylglycine, r-9h-fluoren-9-ylmethoxy carbonyl amino phenyl acetic acid, 2r-2-9h-fluoren-9-yl methoxy carbonyl amino-2-phenylacetic acid, r-9h-fluoren-9-ylmethoxycarbonylamino-phenyl-acetic acid, fmoc-d-alpha-phenylglycine, fmoc-l-, a-phenylglycine
(2R)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-2-phenylacetic acid

Specifications

N-Fmoc-D-alpha-phenylglycine
C23H19NO4
5309006
PCJHOCNJLMFYCV-OAQYLSRUSA-N
(2R)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-2-phenylacetic acid
2756108
Solid
5g
111524-95-9
MFCD00190891
fmoc-d-phg-oh, 2r-2-9h-fluoren-9-ylmethoxycarbonylamino-2-phenyl-acetic acid, r-2-9h-fluoren-9-yl methoxy carbonyl amino-2-phenylacetic acid, fmoc-d-phenylglycine, n-alpha-9-fluorenylmethyloxycarbonyl-d-phenylglycine, r-9h-fluoren-9-ylmethoxy carbonyl amino phenyl acetic acid, 2r-2-9h-fluoren-9-yl methoxy carbonyl amino-2-phenylacetic acid, r-9h-fluoren-9-ylmethoxycarbonylamino-phenyl-acetic acid, fmoc-d-alpha-phenylglycine, fmoc-l-, a-phenylglycine
C1=CC=C(C=C1)C(C(=O)O)NC(=O)OCC2C3=CC=CC=C3C4=CC=CC=C24
373.408
373.4
98%
Videos
SDS