Läs mer
N-Fmoc-D,L-Gamma-carboxyglutamic Acid Gamma,Gamma-Di-t-butyl Ester, TRC
Produktkod. 30403918
Change view
Kvantitet:
1 g
Produktkod. Kvantitet
30403918 1 g
1 options
Produktkod. 30403918 Leverantör Toronto Research Chemicals Leverantörsnummer TRCF6250001G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material N-Fmoc-D,L-gamma-carboxyglutamic Acid gamma,gamma-Di-t-butyl Ester
CAS 308357-14-4
Molekylformel C29 H35 N O8
Kvantitet 1 g
Synonym N-Fmoc-D,L--Carboxyglutamic Acid ,-Di-T-Butyl Ester
InChI formel InChI=1S/C29H35NO8/c1-28(2,3)37-25(33)21(26(34)38-29(4,5)6)15-23(24(31)32)30-27(35)36-16-22-19-13-9-7-11-17(19)18-12-8-10-14-20(18)22/h7-14,21-23H,15-16H2,1-6H3,(H,30,35)(H,31,32)/t23-/m0/s1
LEDER CC(C)(C)OC(=O)C(C[C@H](NC(=O)OCC1c2ccccc2c3ccccc13)C(=O)O)C(=O)OC(C)(C)C
IUPAC-namn (2S)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-5-[(2-methylpropan-2-yl)oxy]-4-[(2-methylpropan-2-yl)oxycarbonyl]-5-oxopentanoic acid
Molekylvikt (g/mol) 525.59
Formel vikt 525.2363