missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-Fmoc-L-methionine sulfone, 98%, Thermo Scientific™
Click to view available options
Quantity:
250 mg
1 g
5 g
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
Color | White |
Synonym | fmoc-met o2-oh, fmoc-l-methionine sulfone, s-2-9h-fluoren-9-yl methoxy carbonyl amino-4-methylsulfonyl butanoic acid, 2s-2-9h-fluoren-9-ylmethoxy carbonyl amino-4-methanesulfonylbutanoic acid, ambotzfaa1406, fmoc-met o 2-oh, pubchem22746, n-alpha-9-fluorenylmethyloxycarbonyl-l-methioninesulfon, s-2-9h-fluoren-9-yl methoxy carbonyl-amino-4-methylsulfonyl butanoic acid, s-2-9h-fluoren-9-yl methoxy carbonylamino-4-methylsulfonyl butanoic acid |
Formula Weight | 403.45 |
Percent Purity | 98% |
Quantity | 5 g |
Physical Form | Powder |
Chemical Name or Material | N-Fmoc-L-methionine sulfone |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'